Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N,N'-Dimethyl-N,N'-bis(3-methylaminopropyl)trimethylenediamine, tech. 90%

Click to view available options
Quantity:
250mg
1g
5g
123-67-1
C13H32N4
244.427
MFCD00014858
XRTKMBBIDHUQNP-UHFFFAOYSA-N
6,10-dimethyl-2,6,10,14-tetraazapentadecane,2,6,10,14-tetraazapentadecane, 6,10-dimethyl,n1,n1'-propane-1,3-diyl bis n1,n3-dimethylpropane-1,3-diamine,n,n'-dimethyl-n,n'-bis 3-methylaminopropyl trimethylenediamine,1,3-propanediamine, n,n'-dimethyl-n,n'-bis 3-methylamino propyl,1,3-propanediamine, n,n'-dimethyl-n,n'-bis 3-methylamino propyl,n,n'-dimethyl-n,n'-bis 3-methylaminopropyl trimeth,1,3-propanediamine, n1,n3-dimethyl-n1,n3-bis 3-methylamino propyl,1,3-propanediamine, n1,n3-dimethyl-n1,n3-bis 3-methylamino propyl,ksc496s7p
67160
N,N'-dimethyl-N'-[3-[methyl-[3-(methylamino)propyl]amino]propyl]propane-1,3-diamine
CNCCCN(C)CCCN(C)CCCNC
123-67-1
C13H32N4
244.427
MFCD00014858
XRTKMBBIDHUQNP-UHFFFAOYSA-N
6,10-dimethyl-2,6,10,14-tetraazapentadecane,2,6,10,14-tetraazapentadecane, 6,10-dimethyl,n1,n1'-propane-1,3-diyl bis n1,n3-dimethylpropane-1,3-diamine,n,n'-dimethyl-n,n'-bis 3-methylaminopropyl trimethylenediamine,1,3-propanediamine, n,n'-dimethyl-n,n'-bis 3-methylamino propyl,1,3-propanediamine, n,n'-dimethyl-n,n'-bis 3-methylamino propyl,n,n'-dimethyl-n,n'-bis 3-methylaminopropyl trimeth,1,3-propanediamine, n1,n3-dimethyl-n1,n3-bis 3-methylamino propyl,1,3-propanediamine, n1,n3-dimethyl-n1,n3-bis 3-methylamino propyl,ksc496s7p
67160
N,N'-dimethyl-N'-[3-[methyl-[3-(methylamino)propyl]amino]propyl]propane-1,3-diamine
CNCCCN(C)CCCN(C)CCCNC
Quantity 5g
Color Yellow
Beilstein 3602172
Formula Weight 244.43
Physical Form Liquid
Percent Purity 90%
Grade Technical
Chemical Name or Material N,N'-Dimethyl-N,N'-bis(3-methylaminopropyl)trimethylenediamine

TSCA : TSCA

storageNote1 : 2° to 8°C