Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethyl-p-phenylenediamine sulfate, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
536-47-0
C8H12N2·H2SO4
234.28
MFCD00012992
GLUKPDKNLKRLHX-UHFFFAOYSA-N
n,n-dimethyl-1,4-phenylenediamine sulfate,n,n-dimethyl-p-phenylenediamine sulfate,n1,n1-dimethylbenzene-1,4-diamine sulfate,1,4-benzenediamine, n,n-dimethyl-, sulfate,unii-x997ivp3ja,4-amino-n,n-dimethylaniline sulphate,x997ivp3ja,1,4-benzenediamine, n,n-dimethyl-, sulfate 1:1,dimethyl-p-phenylenediamine; sulfuric acid,1,4-benzenediamine, n1,n1-dimethyl-, sulfate 1:?
80351
4-N,4-N-dimethylbenzene-1,4-diamine;sulfuric acid
CN(C)C1=CC=C(C=C1)N.OS(=O)(=O)O
536-47-0
C8H12N2·H2SO4
234.28
MFCD00012992
GLUKPDKNLKRLHX-UHFFFAOYSA-N
n,n-dimethyl-1,4-phenylenediamine sulfate,n,n-dimethyl-p-phenylenediamine sulfate,n1,n1-dimethylbenzene-1,4-diamine sulfate,1,4-benzenediamine, n,n-dimethyl-, sulfate,unii-x997ivp3ja,4-amino-n,n-dimethylaniline sulphate,x997ivp3ja,1,4-benzenediamine, n,n-dimethyl-, sulfate 1:1,dimethyl-p-phenylenediamine; sulfuric acid,1,4-benzenediamine, n1,n1-dimethyl-, sulfate 1:?
80351
4-N,4-N-dimethylbenzene-1,4-diamine;sulfuric acid
CN(C)C1=CC=C(C=C1)N.OS(=O)(=O)O
Water 2% max. (K.F.)
Melting Point 234°C to 236°C
Quantity 100g
Boiling Point 495°C
Packaging Glass bottle
Infrared Spectrum Authentic
Color Beige-Gray to Orange or Brown
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (CH3)2NC6H4NH2·H2SO4
Beilstein 13, IV, 106
Formula Weight 234.28
Physical Form Powder
Percent Purity 99%
Chemical Name or Material N, N-Dimethyl-p-phenylenediamine sulfate, 99%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Toxic in contact with skin.
Toxic if swallowed.
Toxic if inhaled.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 208-636-2

TSCA : TSCA