Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N,N-Dimethyl-p-toluidine, 99%

Click to view available options
Quantity:
100g
500g
2500g
99-97-8
C9H13N
135.21
MFCD00008316
GYVGXEWAOAAJEU-UHFFFAOYSA-N
n,n-dimethyl-p-toluidine,dimethyl-p-toluidine,benzenamine, n,n,4-trimethyl,n,n-dimethyl-4-methylaniline,n,n-dimethyl-para-toluidine,4,n,n-trimethylaniline,dimethyl-4-toluidine,p-dimethylamino toluene,n,n-dimethyl-p-tolylamine,n,n,4-trimethylbenzenamine
7471
N,N,4-trimethylaniline
CC1=CC=C(C=C1)N(C)C
99-97-8
C9H13N
135.21
MFCD00008316
GYVGXEWAOAAJEU-UHFFFAOYSA-N
n,n-dimethyl-p-toluidine,dimethyl-p-toluidine,benzenamine, n,n,4-trimethyl,n,n-dimethyl-4-methylaniline,n,n-dimethyl-para-toluidine,4,n,n-trimethylaniline,dimethyl-4-toluidine,p-dimethylamino toluene,n,n-dimethyl-p-tolylamine,n,n,4-trimethylbenzenamine
7471
N,N,4-trimethylaniline
CC1=CC=C(C=C1)N(C)C
Applications
N,N-Dimethyl-p-toluidine is used as a polymerization catalyst for polyesters, acrylate and epoxy resins. It is also used as a hardener for dental cements and in adhesives. It serves as an intermediate for photographic chemicals, in industrial glues, in artificial fingernail preparations, colorants, pharmaceuticals. It reacts with vinyl ether in the presence of copper(II) chloride gives tetrahydroquinolines. Further, it is used to accelerate polymerization of ethyl methacrylate.

Solubility
Miscible with alcohol, ether and chloroform. Immiscible with water.

Notes
Incompatible with strong oxidizing agents. Store in a cool place.
Refractive Index 1.546
Density 0.937
Melting Point -25°C
Flash Point 83°C (181°F)
Boiling Point 210°C to 211°C
Quantity 100g
UN Number UN1708
Beilstein 774409
Solubility Information Miscible with alcohol,ether and chloroform. Immiscible with water.
Formula Weight 135.21
Percent Purity 99%
Chemical Name or Material N,N-Dimethyl-p-toluidine
Show More Show Less

GHS H Statement
H301-H311-H331-H373-H227
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.

P210-P235-P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P370+P378q-P501c

H227-H301+H311+H331-H373

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 202-805-4

RTECSNumber : XU5803000

TSCA : Yes

Recommended Storage : Ambient temperatures