Learn More
N,N-Dimethyl-p-toluidine, 99%, Thermo Scientific Chemicals
Description
N,N-Dimethyl-p-toluidine is used as a polymerization catalyst for polyesters, acrylate and epoxy resins. It is also used as a hardener for dental cements and in adhesives. It serves as an intermediate for photographic chemicals, in industrial glues, in artificial fingernail preparations, colorants, pharmaceuticals. It reacts with vinyl ether in the presence of copper(II) chloride gives tetrahydroquinolines. Further, it is used to accelerate polymerization of ethyl methacrylate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
N,N-Dimethyl-p-toluidine is used as a polymerization catalyst for polyesters, acrylate and epoxy resins. It is also used as a hardener for dental cements and in adhesives. It serves as an intermediate for photographic chemicals, in industrial glues, in artificial fingernail preparations, colorants, pharmaceuticals. It reacts with vinyl ether in the presence of copper(II) chloride gives tetrahydroquinolines. Further, it is used to accelerate polymerization of ethyl methacrylate.
Solubility
Miscible with alcohol, ether and chloroform. Immiscible with water.
Notes
Incompatible with strong oxidizing agents. Store in a cool place.
Specifications
Melting Point | -25°C |
Density | 0.937 |
Boiling Point | 210°C to 211°C |
Flash Point | 83°C (181°F) |
Refractive Index | 1.546 |
Quantity | 100 g |
UN Number | UN1708 |
Beilstein | 774409 |
Solubility Information | Miscible with alcohol,ether and chloroform. Immiscible with water. |
Formula Weight | 135.21 |
Show More |
Safety and Handling
Hazard Category | H227-H301+H311+H331-H373 |
Hazard Statement | GHS H Statement H301-H311-H331-H373-H227 Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. Combustible liquid. |
Precautionary Statement | P210-P235-P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P370+P378q-P501c |
DOTInformation | Hazard Class: 6.1; Packaging Group: III |
EINECSNumber | 202-805-4 |
RTECSNumber | XU5803000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.