Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylacetamide, 99.5%, extra pure, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
20L
Packaging:
Glass Bottle
Metal drum
127-19-5
C4H9NO
87.122
MFCD00008686
FXHOOIRPVKKKFG-UHFFFAOYSA-N
dimethylacetamide,dmac,acetamide, n,n-dimethyl,acetdimethylamide,dimethyl acetamide,n,n-dimethyl acetamide,dimethylamide acetate,n,n-dimethylethanamide,dimethylacetone amide,acetyldimethylamine
31374
CHEBI:84254
N,N-dimethylacetamide
CC(=O)N(C)C
127-19-5
C4H9NO
87.122
MFCD00008686
FXHOOIRPVKKKFG-UHFFFAOYSA-N
dimethylacetamide,dmac,acetamide, n,n-dimethyl,acetdimethylamide,dimethyl acetamide,n,n-dimethyl acetamide,dimethylamide acetate,n,n-dimethylethanamide,dimethylacetone amide,acetyldimethylamine
31374
CHEBI:84254
N,N-dimethylacetamide
CC(=O)N(C)C
CAS Min % 99.4
CAS Max % 100.0
Assay Percent Range 99.5%
Linear Formula CH3CON(CH3)2
Merck Index 15, 3248
Formula Weight 87.12
Physical Form Liquid
Percent Purity 99.5%
Grade Extra Pure
Density 0.9370g/mL
Free Acid 0.1% max.
Infrared Spectrum Authentic
Packaging Metal drum
Refractive Index 1.4370 to 1.4390
Solubility Solubility in water: soluble. Other solubilities: soluble in acetone, diethyl ether, benzene,, chloroform, ethanol and toluene
Specific Gravity 0.937
Water 0.1% max.
Flash Point 70°C
Viscosity 1.02 mPa.s (20°C)
Boiling Point 164.0°C to 166.0°C
Color Undesignated
Melting Point -20.0°C
Quantity 20L
Chemical Name or Material N, N-Dimethylacetamide
Show More Show Less

GHS P Statement
May damage the unborn child.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger