Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylacetamide, 99+%, extra pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
1L
2.5L
5L
25L
Packaging:
Glass Bottle
Metal drum
Plastic drum
127-19-5
C4H9NO
87.122
MFCD00008686
FXHOOIRPVKKKFG-UHFFFAOYSA-N
dimethylacetamide,dmac,acetamide, n,n-dimethyl,acetdimethylamide,dimethyl acetamide,n,n-dimethyl acetamide,dimethylamide acetate,n,n-dimethylethanamide,dimethylacetone amide,acetyldimethylamine
31374
CHEBI:84254
N,N-dimethylacetamide
CC(=O)N(C)C
127-19-5
C4H9NO
87.122
MFCD00008686
FXHOOIRPVKKKFG-UHFFFAOYSA-N
dimethylacetamide,dmac,acetamide, n,n-dimethyl,acetdimethylamide,dimethyl acetamide,n,n-dimethyl acetamide,dimethylamide acetate,n,n-dimethylethanamide,dimethylacetone amide,acetyldimethylamine
31374
CHEBI:84254
N,N-dimethylacetamide
CC(=O)N(C)C

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

CAS Min % 99.0
CAS Max % 100.0
Additional Information Vapor Pressure: 1.7mbar at 25°C
Assay Percent Range 99+%
Linear Formula CH3CON(CH3)2
Beilstein 04, 59
Fieser 01,270; 02,144; 04,165
Merck Index 15, 3248
Formula Weight 87.12
Physical Form Liquid
Percent Purity ≥99.0%
Grade Extra Pure
Density 0.9370g/mL
Free Acid 0.1% max.
Infrared Spectrum Authentic
Packaging Glass Bottle
Refractive Index 1.4370 to 1.4390 (20°C, 589nm)
Residue after Evaporation 0.01% max.
Water 0.1% max.
Color Undesignated
Quantity 1L
Chemical Name or Material N, N-Dimethylacetamide
Show More Show Less

GHS P Statement
May damage the unborn child.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.

Emergency Overview
Combustible liquid. Harmful in contact with skin. Harmful by inhalation. Irritating to eyes. May cause harm to the unborn child.
NFPA
Health:2 Flammability:2 Instability:0

Signal Word: WARNING!