Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylacetamide, 99+%, extra pure, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
5L
25L
Packaging:
Glass Bottle
Metal drum
Plastic drum
127-19-5
C4H9NO
87.122
MFCD00008686
FXHOOIRPVKKKFG-UHFFFAOYSA-N
dimethylacetamide,dmac,acetamide, n,n-dimethyl,acetdimethylamide,dimethyl acetamide,n,n-dimethyl acetamide,dimethylamide acetate,n,n-dimethylethanamide,dimethylacetone amide,acetyldimethylamine
31374
CHEBI:84254
N,N-dimethylacetamide
CC(=O)N(C)C
127-19-5
C4H9NO
87.122
MFCD00008686
FXHOOIRPVKKKFG-UHFFFAOYSA-N
dimethylacetamide,dmac,acetamide, n,n-dimethyl,acetdimethylamide,dimethyl acetamide,n,n-dimethyl acetamide,dimethylamide acetate,n,n-dimethylethanamide,dimethylacetone amide,acetyldimethylamine
31374
CHEBI:84254
N,N-dimethylacetamide
CC(=O)N(C)C
CAS Min % 99.0
CAS Max % 100.0
Additional Information Vapor Pressure: 1.7mbar at 25°C
Assay Percent Range 99+%
Linear Formula CH3CON(CH3)2
Beilstein 04, 59
Fieser 01,270; 02,144; 04,165
Merck Index 15, 3248
Formula Weight 87.12
Physical Form Liquid
Percent Purity ≥99%
Grade Extra Pure
Density 0.9370g/mL
Free Acid 0.1% max.
Infrared Spectrum Authentic
Packaging Plastic drum
Refractive Index 1.4370 to 1.4390
Residue after Evaporation 0.01% max.
Solubility Solubility in water: soluble. Other solubilities: soluble in acetone, diethyl ether, benzene,, chloroform, ethanol and toluene
Specific Gravity 0.937
Water 0.1% max.
Flash Point 70°C
Viscosity 1.02 mPa.s (20°C)
Boiling Point 164.0°C to 166.0°C
Color Undesignated
Melting Point -20.0°C
Quantity 5L
Chemical Name or Material N, N-Dimethylacetamide
Show More Show Less

GHS P Statement
May damage the unborn child.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.

Emergency Overview
Combustible liquid. Harmful in contact with skin. Harmful by inhalation. Irritating to eyes. May cause harm to the unborn child.
NFPA
Health:2 Flammability:2 Instability:0

Signal Word: WARNING!