Learn More
N,N-Dimethylacetamide, anhydrous, 99.8%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Description
N,N-Dimethylacetamide is used as a polar solvent in organic synthesis as well as in reactions involving strong bases such as sodium hydroxide. It is also used as a solvent for fibers like polyacrylonitrile, spandex, as an excipient in drugs viz. vumon, busulfex and in the adhesive industry. It finds application as a reaction medium in the production of pharmaceuticals, agrochemicals, dyes and plasticizers. It is widely used in polymer industry due to its high solving power for high molecular-weight polymers and synthetic resins. It plays an important role as a catalyst in various reactions viz. cyclization, halogenation, cynidation, alkylation and dehydrogenation and increases the yield of main products. Other uses are as extracting agent for oil and gases, in paint removers, in the production of photo-resist stripping compounds and as booster solvent in coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | °C to 20°C |
Density | 0.94 g/mL |
Boiling Point | 165°C to 166°C |
Flash Point | 70°C (158°F) |
Assay Percent Range | 99.8% |
Refractive Index | 1.4385 |
Quantity | 100 mL |
Beilstein | 1737614 |
Solubility Information | Miscible with water,benzene,alcohol,acetone,ether,esters and ketones. |
Formula Weight | 87.12 |
Show More |
Safety and Handling
Hazard Category | H227-H312+H332-H360D |
Precautionary Statement | P201-P202-P210-P261-P271-P281-P302+P352-P304+P340-P308+P313-P312-P363-P370+P378q-P501c |
DOTInformation | Hazard Class: 6.1; Packaging Group: III |
EINECSNumber | 204-826-4 |
RTECSNumber | AB7700000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.