Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylacetamide, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
Narrow Mouth Amber Glass Bottle
127-19-5
C4H9NO
87.122
FXHOOIRPVKKKFG-UHFFFAOYSA-N
31374
CHEBI:84254
N,N-dimethylacetamide
CC(=O)N(C)C
127-19-5
C4H9NO
87.122
FXHOOIRPVKKKFG-UHFFFAOYSA-N
31374
CHEBI:84254
N,N-dimethylacetamide
CC(=O)N(C)C

High quality chemicals for laboratory and specialized industrial use

Formula Weight 87.12
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.94kg
Flash Point 70.00°C
Quantity 4L
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material N,N-Dimethylacetamide