Alfa Aesar™ N,N-Dimethylacetamide-d9, 99% (Isotopic)

Manufacturer: Alfa Aesar™ 4385406

Quantity 5g

 View more versions of this product

Catalog No. AA4385406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 116057-81-9
    Molecular Formula C4D9NO
    Linear Formula CD3CON(CD3)2
    Formula Weight 96.19
    MDL Number MFCD00066061
    Chemical Name or Material N,N-Dimethylacetamide-d9, Isotopic

    Specifications

    Formula Weight 96.19
    Packaging Ampule
    Molecular Formula C4D9NO
    Melting Point −20°C
    Quantity 5g
    Color Colorless
    Flash Point 70°C (158°F)
    Linear Formula CD3CON(CD3)2
    Refractive Index 1.4380
    MDL Number MFCD00066061
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P201-P308+P313
    Obtain special instructions before use.
    IF exposed or concerned: Get medical advice/attention.
    Health Hazard 2 GHS H Statement
    H360-H312-H332-H227
    May damage fertility or the unborn child.
    Harmful in contact with skin.
    Harmful if inhaled.
    Combustible liquid.
    Density 1.035g/mL
    Boiling Point 165°C
    Physical Form Liquid
    CAS 116057-81-9
    Storage Note 1
    • Ampouled under argon
    Chemical Name or Material N,N-Dimethylacetamide-d9, Isotopic
    Assay 99%

    GHS H Statement
    H360-H312-H332-H227
    May damage fertility or the unborn child.
    Harmful in contact with skin.
    Harmful if inhaled.
    Combustible liquid.

    GHS P Statement
    P201-P308+P313
    Obtain special instructions before use.
    IF exposed or concerned: Get medical advice/attention.

    • Ampouled under argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit