Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N,N-Dimethylacetamide-d9, 99% (Isotopic)

Manufacturer:  Alfa Aesar™ 4385403

Quantity 1g

 View more versions of this product

Catalog No. AA4385403


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

116057-81-9
CD3CON(CD3)2
MFCD00066061
C4D9NO
96.19
N,N-Dimethylacetamide-d9, Isotopic

Specifications

1.035g/mL
Ampule
165°C
−20°C
116057-81-9
C4D9NO
96.19
N,N-Dimethylacetamide-d9, Isotopic
70°C (158°F)
1.4380
Colorless
1g
Liquid
CD3CON(CD3)2
MFCD00066061
99%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H360-H312-H332-H227
May damage fertility or the unborn child.
Harmful in contact with skin.
Harmful if inhaled.
Combustible liquid.

GHS P Statement
P201-P308+P313
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

Danger

storageNote1 :

  • Ampouled under argon

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit