Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylacetamide, MilliporeSigma™

Click to view available options
Quantity:
1L
Packaging:
Glass Bottle
127-19-5
C4H9NO
87.12
MFCD00008686
FXHOOIRPVKKKFG-UHFFFAOYSA-N
dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
31374
CHEBI:84254
N,N-dimethylacetamide
CN(C)C(C)=O
127-19-5
C4H9NO
87.12
MFCD00008686
FXHOOIRPVKKKFG-UHFFFAOYSA-N
dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
31374
CHEBI:84254
N,N-dimethylacetamide
CN(C)C(C)=O

For headspace gas chromatography SupraSolv™

Solubility Information Soluble in water at 20°C
Formula Weight 87.12
Physical Form Liquid
Description SupraSolv™ solvents for headspace gas chromatography are developed particularly for the analysis of residual solvents in drug substances, excipients and drug products according to ICH, Ph Eur and USP - SupraSolv headspace solvents are tested for all residual solvents defined by the ICH.
Density 0.94g/cm3 at 20°C
Ignition Point 400°C (752°F)
Odor Amine-like
Packaging Glass Bottle
Refractive Index 1.423
Vapor Pressure 1.76 hPa (20°C)
Flash Point 64°C (147.2°F)
Boiling Point 165°C to 166°C
Color Colorless
Melting Point -20°C
pH 4 (at 200g/L 20°C)
Quantity 1L
Chemical Name or Material N,N-Dimethylacetamide
Show More Show Less

H227 Combustible liquid.

H312 + H332 Harmful in contact with skin or if inhaled.

H360 May damage fertility or the unborn child.