Learn More
N,N-Dimethyldodecylamine, 90+%, Thermo Scientific Chemicals
Description
N,N-Dimethyldodecylamine is used as a as a capping agent during the synthesis of Au(core) -Pd(shell) bimetallic nanoparticles in toluene. It is used in the synthesis of N-alkyl-N,N-dimethyl-1-ammonio-2-hydroxy-3-propane sulfonate. Also used in the preparation of novel bis-quaternary ammonium salt.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
N,N-Dimethyldodecylamine is used as a as a capping agent during the synthesis of Au(core) -Pd(shell) bimetallic nanoparticles in toluene. It is used in the synthesis of N-alkyl-N,N-dimethyl-1-ammonio-2-hydroxy-3-propane sulfonate. Also used in the preparation of novel bis-quaternary ammonium salt.
Solubility
Slightly soluble in water.
Notes
Air & moisture sensitive. Materials to avoid is copper, oxidizing agents, strong acids.
Specifications
Melting Point | -20°C |
Density | 0.787 |
Boiling Point | 110°C to 112°C (3 mmHg) |
Flash Point | 118°C (244°F) |
Refractive Index | 1.4375 |
Quantity | 50 g |
UN Number | UN2735 |
Sensitivity | Air and moisture sensitive |
Solubility Information | Slightly soluble in water. |
Formula Weight | 213.41 |
Show More |
Safety and Handling
Hazard Category | H302-H314 |
Hazard Statement | GHS H Statement H301-H314-H318 Toxic if swallowed. Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
DOTInformation | Hazard Class: 8; Packaging Group: II |
EINECSNumber | 203-943-8 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.