Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Dimethylethylenediamine, tech., 85%, Thermo Scientific Chemicals

Catalog Number p-117590
Click to view available options
Quantity:
5 mL
25 g
100 g
Packaging:
Glass bottle
110-70-3
KVKFRMCSXWQSNT-UHFFFAOYSA-N
n,n'-dimethylethylenediamine, 1,2-bis methylamino ethane, n1,n2-dimethylethane-1,2-diamine, n,n'-dimethyl-1,2-ethanediamine, dimethylethylenediamine, sym-dimethylethylenediamine, 1,2-ethanediamine, n,n'-dimethyl, 2,5-diazahexane, n,n'-dimethyldiaminoethane, n,n'-dimethylethanediamine
8070
N,N'-dimethylethane-1,2-diamine
CNCCNC
110-70-3
KVKFRMCSXWQSNT-UHFFFAOYSA-N
n,n'-dimethylethylenediamine, 1,2-bis methylamino ethane, n1,n2-dimethylethane-1,2-diamine, n,n'-dimethyl-1,2-ethanediamine, dimethylethylenediamine, sym-dimethylethylenediamine, 1,2-ethanediamine, n,n'-dimethyl, 2,5-diazahexane, n,n'-dimethyldiaminoethane, n,n'-dimethylethanediamine
8070
N,N'-dimethylethane-1,2-diamine
CNCCNC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 0.8190g/mL
Boiling Point 119.0°C
Flash Point 26°C
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C4H12N2
Refractive Index 1.4295
Linear Formula CH3NHCH2CH2NHCH3
Quantity 25 g
Beilstein 04, 250
Specific Gravity 0.819
Solubility Information Solubility in water: miscible. Other solubilities: soluble in organic solvents
Molecular Weight (g/mol) 88.15
Formula Weight 88.15
Percent Purity 85%
Grade Technical
Physical Form Liquid
Chemical Name or Material N,N′-Dimethylethylenediamine, tech., 85%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse
EINECSNumber 203-793-3
RTECSNumber KV4250000
TSCA TSCA

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit