Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylformamide, 99.5%, for analysis, ACROS Organics™

Click to view available options
Quantity:
500mL
1L
2.5L
10L
25L
Packaging:
Glass bottle
Plastic drum
68-12-2
C3H7NO
73.10
MFCD00003284
ZMXDDKWLCZADIW-UHFFFAOYSA-N
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
6228
CHEBI:17741
N,N-dimethylformamide
CN(C)C=O
68-12-2
C3H7NO
73.10
MFCD00003284
ZMXDDKWLCZADIW-UHFFFAOYSA-N
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
6228
CHEBI:17741
N,N-dimethylformamide
CN(C)C=O
For analysis
CAS Min % 99.4
CAS Max % 100.0
Acidity Alkalinity 0.0005 meq/g max.
Assay Percent Range 99.5%
Linear Formula HCON(CH3)2
Beilstein 04, 58
Fieser 01,278; 02,153; 03,115; 04,184; 05,247; 07,124; 08,189; 09,182; 11,198; 12,203; 13,124; 14,148; 16,144
Merck Index 15, 3267
Solubility Information Solubility in water: soluble. Other solubilities: miscible with most organic solvents
Calcium (Ca) 1ppm max.
Formula Weight 73.09
Physical Form Liquid
Percent Purity 99.5%
Grade Analysis
Copper (Cu) 0.5ppm max.
Density 0.9450g/mL
Infrared Spectrum Authentic
Iron (Fe) 1ppm max.
Lead (Pb) 0.5ppm max.
Magnesium (Mg) 0.5ppm max.
Packaging Plastic drum
Potassium (K) 0.5ppm max.
Refractive Index 1.4290 to 1.4320
Residue after Evaporation 20ppm max.
Silicon (Si) 0.05ppm max.
Solubility Solubility in water: soluble. Other solubilities: miscible with most organic solvents
Sodium (Na) 2ppm max.
Specific Gravity 0.945
Specific Rotation 0.94
Total Phosphorus 0.5ppm max.
Total Sulfur 1ppm max.
Water 0.05% max.
Zinc (Zn) 0.5ppm max.
Flash Point 58°C
Viscosity 0.8 mPa.s (20°C)
Boiling Point 153.0°C
Color Colorless
Melting Point -61.0°C
Quantity 10L
Chemical Name or Material N,N-Dimethylformamide
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
May damage the unborn child.
Flammable liquid and vapour.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger