Learn More
N,N-Dimethylformamide, 99.5%, for analysis, ACROS Organics™
Description
Specifications
CAS Min % | 99.4 |
CAS Max % | 100.0 |
Acidity Alkalinity | 0.0005 meq/g max. |
Assay Percent Range | 99.5% |
Linear Formula | HCON(CH3)2 |
Beilstein | 04, 58 |
Fieser | 01,278; 02,153; 03,115; 04,184; 05,247; 07,124; 08,189; 09,182; 11,198; 12,203; 13,124; 14,148; 16,144 |
Merck Index | 15, 3267 |
Solubility Information | Solubility in water: soluble. Other solubilities: miscible with most organic solvents |
Calcium (Ca) | 1ppm max. |
Show More |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
May damage the unborn child.
Flammable liquid and vapour.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger