Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylformamide, 99+%, extra pure, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
5L
25L
Packaging:
Glass Bottle
Plastic drum
68-12-2
C3H7NO
73.095
MFCD00003284
ZMXDDKWLCZADIW-UHFFFAOYSA-N
dimethylformamide,dimethyl formamide,n,n-dimethylmethanamide,n-formyldimethylamine,formamide, n,n-dimethyl,dmf,dimethylformamid,dimetilformamide,dwumetyloformamid,dmfa
6228
CHEBI:17741
N,N-dimethylformamide
CN(C)C=O
68-12-2
C3H7NO
73.095
MFCD00003284
ZMXDDKWLCZADIW-UHFFFAOYSA-N
dimethylformamide,dimethyl formamide,n,n-dimethylmethanamide,n-formyldimethylamine,formamide, n,n-dimethyl,dmf,dimethylformamid,dimetilformamide,dwumetyloformamid,dmfa
6228
CHEBI:17741
N,N-dimethylformamide
CN(C)C=O
CAS Min % 99.0
CAS Max % 100.0
Additional Information Vapor Pressure: 4.9mbar at 20°C
Assay Percent Range 99+%
Linear Formula HCON(CH3)2
Beilstein 04, 58
Fieser 01,278; 02,153; 03,115; 04,184; 05,247; 07,124; 08,189; 09,182; 11,198; 12,203; 13,124; 14,148; 16,144
Merck Index 15, 3267
Solubility Information Solubility in water: soluble. Other solubilities: miscible with most organic solvents
Formula Weight 73.09
Physical Form Liquid
Percent Purity ≥99%
Grade Extra Pure
Density 0.9450g/mL
Packaging Glass Bottle
Refractive Index 1.4290 to 1.4320
Residue after Evaporation 0.005% max.
Specific Gravity 0.945
Specific Rotation 0.94
Water 0.2% max. (K.F.)
Flash Point 58°C
Viscosity 0.8 mPa.s (20°C)
Decomposition Information >350°C
Boiling Point 153.0°C
Color Undesignated
Melting Point -61.0°C
Quantity 1L
Chemical Name or Material N,N-Dimethylformamide, 99%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
May damage the unborn child.
Flammable liquid and vapour.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 200-679-5

RTECSNumber : LQ2100000

TSCA : TSCA