Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylformamide, Alfa Aesar™

Click to view available options
Quantity:
500mL
1L
4 x 1L
68-12-2
C3H7NO
73.095
MFCD00003284
ZMXDDKWLCZADIW-UHFFFAOYSA-N
dimethylformamide,dimethyl formamide,n,n-dimethylmethanamide,n-formyldimethylamine,formamide, n,n-dimethyl,dmf,dimethylformamid,dimetilformamide,dwumetyloformamid,dmfa
6228
CHEBI:17741
N,N-dimethylformamide
CN(C)C=O
68-12-2
C3H7NO
73.095
MFCD00003284
ZMXDDKWLCZADIW-UHFFFAOYSA-N
dimethylformamide,dimethyl formamide,n,n-dimethylmethanamide,n-formyldimethylamine,formamide, n,n-dimethyl,dmf,dimethylformamid,dimetilformamide,dwumetyloformamid,dmfa
6228
CHEBI:17741
N,N-dimethylformamide
CN(C)C=O

A solvent For Chromogenic Substrates

Maximum level of impurities
  • Appearance Clear
  • Color (APHA) 15
  • Evaporation residue 0.005%
  • Titratable base 0.003meq/g
  • Titratable acid 0.0005meq/g
  • H2O 0.15%
Flash Point 57°C (135°F)
Boiling Point 153°C
Refractive Index 1.431
Melting Point °C to 61°C
Density 0.944 g/cm3 at 20°C
Appearance Colorless liquid
Odor Amine-like
Vapor Pressure 3.5 hPa (3mm Hg) at 20°C
Color Colorless
Quantity 500mL
Ignition Point 440°C (824°F)
Assay Percent Range >99.8%
Beilstein 605365
Solubility Information Miscible with water and most organic solvents.
Formula Weight 73.09
Physical Form Liquid
Grade ACS
Chemical Name or Material N, N-Dimethylformamide, For analysis ACS
Show More Show Less

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P312-P363-P370+P378q-P501c

H226-H312+H332-H319-H335-H336-H350-H360D

DOTInformation : DOT Class: 3, Packaging Group: III

EINECSNumber : 200-679-5

RTECSNumber : LQ2100000

TSCA : Yes

Recommended Storage : Ambient temperatures