N,N-Dimethylformamide, anhydrous, 99.8%, Alfa Aesar™

Manufacturer: Alfa Aesar 43997AE

Quantity 100mL

 View more versions of this product

Catalog No. AA43997AE

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 68-12-2
    Physical Form Liquid
    Assay Percent Range 99.8%
    Molecular Formula HCON(CH3)2
    Formula Weight 73.09
    MDL Number MFCD00003284
    Chemical Name or Material N, N-Dimethylformamide

    Specifications

    Color Clear colorless
    Formula Weight 73.09
    Physical Form Liquid
    Quantity 100mL
    UN Number 2265
    DOT Information DOT Class: 3, Packaging Group: III
    Density 0.944g/cm3 at 20°C
    EINECS Number 200-679-5
    Odor Amine-like
    Ignition Point 440°C (824°F)
    Vapor Pressure 3mm Hg at 20°C
    Assay Percent Range 99.8%
    Molecular Formula HCON(CH3)2
    MDL Number MFCD00003284
    CAS 68-12-2
    Health Hazard 1 H360-H226-H312-H332-H319: May damage fertility or the unborn child. Flammable liquid and vapour. Harmful in contact with skin. Harmful if inhaled. Causes serious eye irritation.
    Chemical Name or Material N, N-Dimethylformamide

    H360-H226-H312-H332-H319: May damage fertility or the unborn child. Flammable liquid and vapour. Harmful in contact with skin. Harmful if inhaled. Causes serious eye irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit