Alfa Aesar™ N,N-Dimethylformamide dibenzyl acetal, tech. 80%

Manufacturer: Alfa Aesar™ A1128130

Quantity 250g

 View more versions of this product

Catalog No. AAA1128130

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2016-04-8
    Molecular Formula (CH3)2NCH(OCH2C6H5)2
    Formula Weight 271.36
    MDL Number MFCD00014869
    Chemical Name or Material N,N-Dimethylformamide dibenzyl acetal
    Grade Technical Grade

    Specifications

    Formula Weight 271.36
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 2016-04-8
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Quantity 250g
    Molecular Formula (CH3)2NCH(OCH2C6H5)2
    MDL Number MFCD00014869
    Health Hazard 1 Warning
    Boiling Point 138° to 140°C (0.5mmHg)
    Chemical Name or Material N,N-Dimethylformamide dibenzyl acetal
    Assay 85%
    Grade Technical Grade

    Description

    Applications
    N,N-Dimethylformamide diethyl acetal was used in quantification of cocaine and its primary metabolite, benzoyl ecgonine from urine matrix by gas chromatography.

    Solubility
    Soluble in variety of inert solvents.

    Notes
    Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit