N,N-Dimethylformamide, Spectrophotometric Grade, >99.7%, Alfa Aesar™

Maximum level of impurities: Evaporation residue 5ppm, H2O 0.1%

Manufacturer: Alfa Aesar 13808K2

Quantity 1L

 View more versions of this product

Catalog No. AA13808K2

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 68-12-2
    Physical Form Liquid
    Assay Percent Range >99.7%
    Molecular Formula HCON(CH3)2
    Formula Weight 73.09
    MDL Number MFCD00003284
    Chemical Name or Material N, N-Dimethylformamide, Maximum level of impurities: Evaporation residue 5ppm, H2O 0.1%
    Grade Spectrophotometric grade

    Specifications

    Physical Form Liquid
    Formula Weight 73.09
    Color Clear colorless
    Quantity 1L
    DOT Information Hazard Class: 3, Packing Group: III
    Density 0.944g/cm3
    EINECS Number 200-679-5
    Ignition Point 440°C (824°F)
    Assay Percent Range >99.7%
    MDL Number MFCD00003284
    CAS 68-12-2
    Vapor Pressure 3.5 hPa (3mm Hg) at 20°C
    Odor Amine-like
    Molecular Formula HCON(CH3)2
    Health Hazard 1 H360-H226-H312-H332-H319: May damage fertility or the unborn child. Flammable liquid and vapour. Harmful in contact with skin. Harmful if inhaled. Causes serious eye irritation.
    Appearance Colorless liquid
    Chemical Name or Material N, N-Dimethylformamide, Maximum level of impurities: Evaporation residue 5ppm, H2O 0.1%
    Grade Spectrophotometric grade

    H360-H226-H312-H332-H319: May damage fertility or the unborn child. Flammable liquid and vapour. Harmful in contact with skin. Harmful if inhaled. Causes serious eye irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit