Alfa Aesar™ N,N-Dimethylhexanamide, 98%

Manufacturer: Alfa Aesar™ B2456206

Quantity 5g

 View more versions of this product

Catalog No. AAB2456206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5830-30-8
    Molecular Formula CH3(CH2)4CON(CH3)2
    Formula Weight 143.23
    MDL Number MFCD00043671
    Beilstein 1747074
    Synonym N,N-Dimethylcaproamide
    Chemical Name or Material N,N-Dimethylhexanamide

    Specifications

    Formula Weight 143.23
    TSCA TSCA
    Quantity 5g
    Molecular Formula CH3(CH2)4CON(CH3)2
    MDL Number MFCD00043671
    Synonym N,N-Dimethylcaproamide
    Health Hazard 2 GHS H Statement
    H302
    Harmful if swallowed.
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number MN7950000
    Boiling Point 80°C (1mmHg)
    CAS 5830-30-8
    Beilstein 1747074
    Health Hazard 1 Warning
    Chemical Name or Material N,N-Dimethylhexanamide
    Assay 98%

    Description

    Applications
    N,N-Dimethylhexanamide is used as a pharmaceutical intermediate.

    Solubility
    Not miscible or difficult to mix.

    Notes
    Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

    GHS H Statement
    H302
    Harmful if swallowed.

    GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit