Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylmethyleneiminium iodide, 97%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
33797-51-2
C3H8IN
185.01
MFCD00011810
VVDUZZGYBOWDSQ-UHFFFAOYSA-M
eschenmoser's salt, n,n-dimethylmethyleneiminium iodide, n-methyl-n-methylenemethanaminium iodide, dimethyl methylidene azanium iodide, n,n-dimethyl methyleneammonium iodide, dimethyl methylene ammonium iodide, methanaminium, n-methyl-n-methylene-, iodide, eschenmoser inverted exclamation mark s salt, eschenmoser salt, dimethyl methylene ammounium iodide
2724133
dimethyl(methylidene)azanium;iodide
C[N+](=C)C.[I-]
33797-51-2
C3H8IN
185.01
MFCD00011810
VVDUZZGYBOWDSQ-UHFFFAOYSA-M
eschenmoser's salt, n,n-dimethylmethyleneiminium iodide, n-methyl-n-methylenemethanaminium iodide, dimethyl methylidene azanium iodide, n,n-dimethyl methyleneammonium iodide, dimethyl methylene ammonium iodide, methanaminium, n-methyl-n-methylene-, iodide, eschenmoser inverted exclamation mark s salt, eschenmoser salt, dimethyl methylene ammounium iodide
2724133
dimethyl(methylidene)azanium;iodide
C[N+](=C)C.[I-]
Melting Point 219°C
Packaging Glass bottle
Color Cream to Yellow
Infrared Spectrum Authentic
Quantity 50g
Assay Percent Range 97%
Linear Formula H2C=N(CH3)2I
Beilstein 04, IV, 153
Fieser 04,186; 08,194; 10,160; 13,121
Merck Index 15, 3275
Formula Weight 185.01
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material N, N-Dimethylmethyleneiminium iodide, 97%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 251-680-2