Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N,N-Dimethylthioformamide, 97%

Click to view available options
Quantity:
5g
25g
100g
758-16-7
C3H7NS
89.156
MFCD00004943
SKECXRFZFFAANN-UHFFFAOYSA-N
n,n-dimethylthioformamide,dimethylthioformamide,methanethioamide, n,n-dimethyl,formamide, n,n-dimethylthio,nn-dimethylthioformamide,methanethioamide,n,n-dimethyl,formamide,n-dimethylthio,dimethylamino methanethial,n,n-dimethyl-thioformamide,methanethioamide, dimethyl
69794
N,N-dimethylmethanethioamide
CN(C)C=S
758-16-7
C3H7NS
89.156
MFCD00004943
SKECXRFZFFAANN-UHFFFAOYSA-N
n,n-dimethylthioformamide,dimethylthioformamide,methanethioamide, n,n-dimethyl,formamide, n,n-dimethylthio,nn-dimethylthioformamide,methanethioamide,n,n-dimethyl,formamide,n-dimethylthio,dimethylamino methanethial,n,n-dimethyl-thioformamide,methanethioamide, dimethyl
69794
N,N-dimethylmethanethioamide
CN(C)C=S
Applications
It is used in telecommunication.

Solubility
Fully miscible in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
Quantity 5g
Boiling Point 58°C to 60°C (1mmHg)
Formula Weight 89.15
Percent Purity 97%
Chemical Name or Material N,N-Dimethylthioformamide

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA