Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Dimethylthiourea, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
Packaging:
Glass bottle
534-13-4
C3H8N2S
104.18
MFCD00004923
VLCDUOXHFNUCKK-UHFFFAOYSA-N
n,n'-dimethylthiourea,1,3-dimethyl-2-thiourea,thiourea, n,n'-dimethyl,sym-dimethylthiourea,dimethylthiocarbamide,dimethylthiourea,1,3-dimethylisothiourea,dimethyl-2-thiourea,urea, 1,3-dimethyl-2-thio,unii-8p30pmd17w
2723631
1,3-dimethylthiourea
CNC(=S)NC
534-13-4
C3H8N2S
104.18
MFCD00004923
VLCDUOXHFNUCKK-UHFFFAOYSA-N
n,n'-dimethylthiourea,1,3-dimethyl-2-thiourea,thiourea, n,n'-dimethyl,sym-dimethylthiourea,dimethylthiocarbamide,dimethylthiourea,1,3-dimethylisothiourea,dimethyl-2-thiourea,urea, 1,3-dimethyl-2-thio,unii-8p30pmd17w
2723631
1,3-dimethylthiourea
CNC(=S)NC
Boiling Point 155°C (1.0 mbar)
Flash Point 200°C
Color Colorless to White
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 61°C to 67°C
Quantity 5g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CH3NHCSNHCH3
Beilstein 04, 70
Merck Index 15, 3287
Solubility Information Solubility in water: 2900g/L (20°C). Other solubilities: soluble in alcohol and acetone,sparingly soluble in benzene,ether and carbon,disulfide,slightly soluble in petroleum ether
Formula Weight 104.18
Physical Form Crystals, Crystalline Powder and/or Chunks
Percent Purity ≥98.5%
Chemical Name or Material N, N'-Dimethylthiourea, 99%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
May cause an allergic skin reaction.

GHS P Statement:
Wear protective gloves/protective clothing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 208-588-2

RTECSNumber : YT1520000

TSCA : TSCA