Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethyltrimethylsilylamine, 97%, ACROS Organics™

Click to view available options
Quantity:
10mL
50mL
Packaging:
Glass bottle
2083-91-2
C5H15NSi
117.27
MFCD00008297
KAHVZNKZQFSBFW-UHFFFAOYSA-N
n,n-dimethyltrimethylsilylamine,n,n-dimethylaminotrimethylsilane,silanamine, pentamethyl,dimethylamino trimethylsilane,pentamethylsilylamine,n-trimethylsilyl dimethylamine,tmsdma,dimethylaminotrimethylsilane,silanamine, n,n,1,1,1-pentamethyl,n,n-dimethyltrimethylsilamine
74965
N-methyl-N-trimethylsilylmethanamine
CN(C)[Si](C)(C)C
2083-91-2
C5H15NSi
117.27
MFCD00008297
KAHVZNKZQFSBFW-UHFFFAOYSA-N
n,n-dimethyltrimethylsilylamine,n,n-dimethylaminotrimethylsilane,silanamine, pentamethyl,dimethylamino trimethylsilane,pentamethylsilylamine,n-trimethylsilyl dimethylamine,tmsdma,dimethylaminotrimethylsilane,silanamine, n,n,1,1,1-pentamethyl,n,n-dimethyltrimethylsilamine
74965
N-methyl-N-trimethylsilylmethanamine
CN(C)[Si](C)(C)C
Refractive Index 1.3960 to 1.398
Boiling Point 84°C
Flash Point -19°C
Specific Gravity 0.732
Infrared Spectrum Authentic
Quantity 10mL
Color Colorless to Yellow
Packaging Glass bottle
Density 0.7320g/mL
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula (CH3)3SiN(CH3)2
Solubility Information Solubility in water: decomposes
Formula Weight 117.27
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material N, N-Dimethyltrimethylsilylamine
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 218-222-3