Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N,N-Dimethyltrimethylsilylamine, 97%, ACROS Organics™
Click to view available options
Quantity:
10mL
50mL
Packaging:
Glass bottle
Specifications
Specific Gravity | 0.732 |
Boiling Point | 84°C |
Flash Point | -19°C |
Refractive Index | 1.3960 to 1.398 |
Density | 0.7320g/mL |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Quantity | 50mL |
Color | Colorless to Yellow |
CAS Min % | 96.0 |
Show More |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 218-222-3