Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Disuccinimidyl carbonate, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
74124-79-1
C9H8N2O7
256.16
MFCD00009767
PFYXSUNOLOJMDX-UHFFFAOYSA-N
n,n'-disuccinimidyl carbonate,bis 2,5-dioxopyrrolidin-1-yl carbonate,n,n'-disuccinimidylcarbonate,di succinimido carbonate,disuccinimidyl carbonate,di n-succinimidyl carbonate,n,n-disuccinimidyl carbonate,dsc,disuccinimido carbonate,pubchem12700
676246
bis(2,5-dioxopyrrolidin-1-yl) carbonate
C1CC(=O)N(C1=O)OC(=O)ON2C(=O)CCC2=O
74124-79-1
C9H8N2O7
256.16
MFCD00009767
PFYXSUNOLOJMDX-UHFFFAOYSA-N
n,n'-disuccinimidyl carbonate,bis 2,5-dioxopyrrolidin-1-yl carbonate,n,n'-disuccinimidylcarbonate,di succinimido carbonate,disuccinimidyl carbonate,di n-succinimidyl carbonate,n,n-disuccinimidyl carbonate,dsc,disuccinimido carbonate,pubchem12700
676246
bis(2,5-dioxopyrrolidin-1-yl) carbonate
C1CC(=O)N(C1=O)OC(=O)ON2C(=O)CCC2=O
Coupling reagent
Elemental Analysis 97.5% min.
Quantity 5g
Color White to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 190.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Fieser 09,208; 11,226; 01,629; 07,218; 13,259; 17,174
Formula Weight 256.16
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material N,N′-Disuccinimidyl carbonate, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 277-730-3