Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-n-Heptylmethylamine, 96%

Click to view available options
Quantity:
5g
25g
36343-05-2
C8H19N
129.247
MFCD00039980
LTGYRKOQQQWWAF-UHFFFAOYSA-N
n-heptylmethylamine,n-methylheptylamine,1-heptanamine, n-methyl,heptylmethylamine,heptyl methyl amine,n-methyl-1-heptanamine,methyl heptylamine,methylheptyl-amine,n-methylaminoheptane,acmc-20ak48
93190
N-methylheptan-1-amine
CCCCCCCNC
36343-05-2
C8H19N
129.247
MFCD00039980
LTGYRKOQQQWWAF-UHFFFAOYSA-N
n-heptylmethylamine,n-methylheptylamine,1-heptanamine, n-methyl,heptylmethylamine,heptyl methyl amine,n-methyl-1-heptanamine,methyl heptylamine,methylheptyl-amine,n-methylaminoheptane,acmc-20ak48
93190
N-methylheptan-1-amine
CCCCCCCNC
Color Colorless
Quantity 5g
Density 0.768g/mL
Boiling Point 53°C to 58°C (6mmHg)
Refractive Index 1.4230
Sensitivity Moisture Sensitive
Linear Formula CH3(CH2)6NHCH3
UN Number UN2734
Beilstein 906719
Formula Weight 129.25
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material N-n-Heptylmethylamine
Show More Show Less

GHS H Statement
H314-H318-H226

GHS P Statement
P210-P260-P280-P303+P361+P353-P305+P351+P338-P301+P330+P331-P304+P340-P310-P405-P501a

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.