Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Hexamethylenebisacetamide, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
50g
250g
Packaging:
Glass bottle
3073-59-4
C10H20N2O2
200.282
MFCD00008684
BNQSTAOJRULKNX-UHFFFAOYSA-N
hmba,hexamethylene bisacetamide,hexamethylenebisacetamide,n,n'-hexamethylenebisacetamide,hexamethylenediacetamide,n,n'-diacetyl-1,6-diaminohexane,diacetyldiaminohexane,n,n'-diacetylhexamethylenediamine,n,n'-hexane-1,6-diyl diacetamide,acetamide, n,n'-hexamethylenebis
3616
N-(6-acetamidohexyl)acetamide
CC(=O)NCCCCCCNC(=O)C
3073-59-4
C10H20N2O2
200.282
MFCD00008684
BNQSTAOJRULKNX-UHFFFAOYSA-N
hmba,hexamethylene bisacetamide,hexamethylenebisacetamide,n,n'-hexamethylenebisacetamide,hexamethylenediacetamide,n,n'-diacetyl-1,6-diaminohexane,diacetyldiaminohexane,n,n'-diacetylhexamethylenediamine,n,n'-hexane-1,6-diyl diacetamide,acetamide, n,n'-hexamethylenebis
3616
N-(6-acetamidohexyl)acetamide
CC(=O)NCCCCCCNC(=O)C
Melting Point 126°C to 129°C
Quantity 50g
Color White
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3CONH(CH2)6NHCOCH3
Beilstein 04, 269
Formula Weight 200.28
Physical Form Crystalline Powder of Flakes
Percent Purity ≥97.5%
Chemical Name or Material N, N'-Hexamethylenebisacetamide
Show More Show Less