Alfa Aesar™ N-n-Hexylaniline, 98%

Manufacturer: Alfa Aesar™ H5552406

Quantity 5g

 View more versions of this product

Catalog No. AAH5552406

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 4746-32-1
    Formula Weight 177.29
    MDL Number MFCD00462723
    Chemical Name or Material N-n-Hexylaniline

    Specifications

    Formula Weight 177.29
    Boiling Point 140° to 145°C (15mmHg)
    Refractive Index 1.5230
    MDL Number MFCD00462723
    Physical Form Liquid
    CAS 4746-32-1
    Density 0.899g/mL
    Color Colorless to brown
    Quantity 5g
    Chemical Name or Material N-n-Hexylaniline
    Assay 98%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit