Alfa Aesar™ N,N,N',N'-Tetraethylmethylenediamine, 98%

Manufacturer: Alfa Aesar™ 1129414

Quantity 25g

 View more versions of this product

Catalog No. AA1129414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 102-53-4
    Formula Weight 158.29
    MDL Number MFCD00015115
    UN Number UN2733
    Synonym Bis(diethylamino)methane
    Chemical Name or Material N,N,N',N'-Tetraethylmethylenediamine

    Specifications

    Formula Weight 158.29
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN2733
    Boiling Point 166° to 169°C
    Flash Point 42° (108°F)
    MDL Number MFCD00015115
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
    Refractive Index 1.4260
    Density 0.811g/mL
    Quantity 25g
    CAS 102-53-4
    Synonym Bis(diethylamino)methane
    TSCA TSCA
    Physical Form Liquid
    Health Hazard 2 GHS H Statement
    H314-H318-H226
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapor.
    Chemical Name or Material N,N,N',N'-Tetraethylmethylenediamine
    Assay 98%

    GHS H Statement
    H314-H318-H226
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapor.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit