Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,N',N'-Tetramethyl-1,3-propanediamine, 99+%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
250g
Packaging:
Glass bottle
110-95-2
C7H18N2
130.23
MFCD00008337
DMQSHEKGGUOYJS-UHFFFAOYSA-N
n,n,n',n'-tetramethyl-1,3-propanediamine,1,3-bis dimethylamino propane,tetramethyltrimethylenediamine,n1,n1,n3,n3-tetramethylpropane-1,3-diamine,n,n,n',n'-tetramethyl-1,3-diaminopropane,1,3-propanediamine, n,n,n',n'-tetramethyl,bis dimethylamino methyl methane,n,n,n',n'-tetramethyltrimethylenediamine,unii-73b9i6hn5r
8084
N,N,N',N'-tetramethylpropane-1,3-diamine
CN(C)CCCN(C)C
110-95-2
C7H18N2
130.23
MFCD00008337
DMQSHEKGGUOYJS-UHFFFAOYSA-N
n,n,n',n'-tetramethyl-1,3-propanediamine,1,3-bis dimethylamino propane,tetramethyltrimethylenediamine,n1,n1,n3,n3-tetramethylpropane-1,3-diamine,n,n,n',n'-tetramethyl-1,3-diaminopropane,1,3-propanediamine, n,n,n',n'-tetramethyl,bis dimethylamino methyl methane,n,n,n',n'-tetramethyltrimethylenediamine,unii-73b9i6hn5r
8084
N,N,N',N'-tetramethylpropane-1,3-diamine
CN(C)CCCN(C)C
Packaging Glass bottle
Specific Gravity 0.78
Infrared Spectrum Authentic
Refractive Index 1.4224 to 1.4244
Flash Point 31°C
Quantity 50g
Melting Point -70.0°C
Density 0.7800g/mL
Color Colorless to Yellow
Boiling Point 145.0°C to 146.0°C
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99% min. (GC)
Linear Formula (CH3)2N(CH2)3N(CH3)2
Beilstein 04, 262
Formula Weight 130.23
Physical Form Liquid
Percent Purity 99+%
Chemical Name or Material N,N,N′,N′-Tetramethyl-1,3-propanediamine
Show More Show Less

GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Toxic if inhaled.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth

GHS Signal Word: Danger

EINECSNumber : 203-818-8