Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,N',N'-Tetramethyl-1,4-butanediamine, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
111-51-3
C8H20N2
144.262
MFCD00008338
VEAZEPMQWHPHAG-UHFFFAOYSA-N
n,n,n',n'-tetramethyl-1,4-butanediamine,tetramethyldiaminobutane,tetramethylputrescine,n1,n1,n4,n4-tetramethylbutane-1,4-diamine,1,4-bis dimethylamino butane,1,4-butanediamine, n,n,n',n'-tetramethyl,n,n,n',n'-tetramethyl-1,4-diaminobutane,n,n,n',n'-tetramethyltetramethylenediamine,unii-5j7765fhlu,alpha,delta-tetramethyldiamino butane
66074
N,N,N',N'-tetramethylbutane-1,4-diamine
CN(C)CCCCN(C)C
111-51-3
C8H20N2
144.262
MFCD00008338
VEAZEPMQWHPHAG-UHFFFAOYSA-N
n,n,n',n'-tetramethyl-1,4-butanediamine,tetramethyldiaminobutane,tetramethylputrescine,n1,n1,n4,n4-tetramethylbutane-1,4-diamine,1,4-bis dimethylamino butane,1,4-butanediamine, n,n,n',n'-tetramethyl,n,n,n',n'-tetramethyl-1,4-diaminobutane,n,n,n',n'-tetramethyltetramethylenediamine,unii-5j7765fhlu,alpha,delta-tetramethyldiamino butane
66074
N,N,N',N'-tetramethylbutane-1,4-diamine
CN(C)CCCCN(C)C
Quantity 5g
Infrared Spectrum Authentic
Refractive Index 1.4270 to 1.4300 (20°C, 589nm)
Density 0.7900g/mL
Color Colorless
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula (CH3)2N(CH2)4N(CH3)2
Beilstein 04, 265
Merck Index 15, 9373
Formula Weight 144.26
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material N,N,N',N'-Tetramethyl-1,4-butanediamine, 97%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-878-5

RTECSNumber : EJ7530000

TSCA : TSCA