Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N,N,N',N'-Tetramethyl-O-(4-oxo-3,4-dihydro-1,2,3-benzotriazin-3-yl)uronium tetrafluoroborate, 98+%

Manufacturer:  Alfa Aesar™ H5414406

Quantity 5g

 View more versions of this product

Catalog No. AAH5414406


  Check Availability
Add to cart

Specifications

Description

Applications
O-(3,4-Dihydro-4-oxo-1,2,3-benzotriazin-3-yl)-N,N,N ft.,N ft.-tetramethyluronium tetrafluoroborate (2-(3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-yl)-1,1,3,3-tetramethyluronium tetrafluoroborate, TBTU) may be used as coupling reagent in the synthesis of macrocyclic polyamine derivatives with various length of linkers, condensation reagent in the large-scale preparation of the hematoregulatory nonapeptide (Glp-Glu-Asp)2-DAS-(Lys)2 (2,SK&F 107647), coupling reagent for peptide synthesis which best suppresses racemization during fragment condensation.

Solubility
Soluble in acetonitrile 0.1 g/mL.

Notes
Moisture and light sensitive. Store away from water/ moisture, oxidizing agents. Protect from heat.
Description & Specifications

Chemical Identifiers

125700-69-8
349.09
O-(3,4-Dihydro-4-oxo-1,2,3-benzotriazin-3-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate; TDBTU
C12H16BF4N5O2
MFCD00077730
N,N,N',N'-Tetramethyl-O-(4-oxo-3,4-dihydro-1,2,3-benzotriazin-3-yl)uronium tetrafluoroborate

Specifications

125700-69-8
349.09
O-(3,4-Dihydro-4-oxo-1,2,3-benzotriazin-3-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate; TDBTU
N,N,N',N'-Tetramethyl-O-(4-oxo-3,4-dihydro-1,2,3-benzotriazin-3-yl)uronium tetrafluoroborate
C12H16BF4N5O2
MFCD00077730
5g
≥98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit