Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,N',N'-Tetramethyl-O-(N-succinimidyl)uronium hexafluorophosphate, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
265651-18-1
C9H16N3O3·F6P
359.21
STWZCCVNXFLDDD-UHFFFAOYSA-N
hstu,n,n,n',n'-tetramethyl-o-n-succinimidyl uronium hexafluorophosphate,o-n-succinimidyl-n,n,n',n'-tetramethyluronium hexafluorophosphate,hstu n,n,n',n'-tetramethyl-o-n-succinimidyl uronium hexafluorophosphate,dimethylamino 2,5-dioxopyrrolidin-1-yl oxy methylidene dimethylazanium hexafluorophosphate,ambotzrl-1039,o-n-succinimidyl-1,1,3,3-tetramethyluronium hexafluorophosphate,2-succinimido-1,1,3,3-tetramethyluronim hexafluorophosphate,2-succinimido-1,1,3,3-tetramethyluronium hexafluorophosphate,n,n,n',n'-tetramethyl-o-n-succinimidyl uroniumhexafluorophosphate
16211151
[dimethylamino-(2,5-dioxopyrrolidin-1-yl)oxymethylidene]-dimethylazanium;hexafluorophosphate
CN(C)C(=[N+](C)C)ON1C(=O)CCC1=O.F[P-](F)(F)(F)(F)F
265651-18-1
C9H16N3O3·F6P
359.21
STWZCCVNXFLDDD-UHFFFAOYSA-N
hstu,n,n,n',n'-tetramethyl-o-n-succinimidyl uronium hexafluorophosphate,o-n-succinimidyl-n,n,n',n'-tetramethyluronium hexafluorophosphate,hstu n,n,n',n'-tetramethyl-o-n-succinimidyl uronium hexafluorophosphate,dimethylamino 2,5-dioxopyrrolidin-1-yl oxy methylidene dimethylazanium hexafluorophosphate,ambotzrl-1039,o-n-succinimidyl-1,1,3,3-tetramethyluronium hexafluorophosphate,2-succinimido-1,1,3,3-tetramethyluronim hexafluorophosphate,2-succinimido-1,1,3,3-tetramethyluronium hexafluorophosphate,n,n,n',n'-tetramethyl-o-n-succinimidyl uroniumhexafluorophosphate
16211151
[dimethylamino-(2,5-dioxopyrrolidin-1-yl)oxymethylidene]-dimethylazanium;hexafluorophosphate
CN(C)C(=[N+](C)C)ON1C(=O)CCC1=O.F[P-](F)(F)(F)(F)F
Coupling reagent
Melting Point 213°C to 215°C
Color White
Elemental Analysis 29.34 to 30.85% C, 11.41 to 11.99% N
Quantity 100g
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 359.21
Physical Form Powder or Crystalline Powder
Percent Purity 98%
Chemical Name or Material N,N,N',N'-Tetramethyl-O-(N-succinimidyl)uronium hexafluorophosphate
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning