Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,N',N'-Tetramethylbenzidine 97.5%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
366-29-0
C16H20N2
240.35
MFCD00008310
YRNWIFYIFSBPAU-UHFFFAOYSA-N
n,n,n',n'-tetramethylbenzidine,benzidine, n,n,n',n'-tetramethyl,4,4'-bis n,n-dimethylamino biphenyl,n,n,n',n'-tetramethyl-p,p'-benzidine,ccris 1000,1,1'-biphenyl-4,4'-diamine, n,n,n',n'-tetramethyl,n,n,n',n'-tetramethyl-1,1'-biphenyl-4,4'-diamine,4-4-dimethylamino phenyl phenyl dimethylamine,1,1'-biphenyl-4,4'-diamine, n4,n4,n4',n4'-tetramethyl
9702
4-[4-(dimethylamino)phenyl]-N,N-dimethylaniline
CN(C)C1=CC=C(C=C1)C2=CC=C(C=C2)N(C)C
366-29-0
C16H20N2
240.35
MFCD00008310
YRNWIFYIFSBPAU-UHFFFAOYSA-N
n,n,n',n'-tetramethylbenzidine,benzidine, n,n,n',n'-tetramethyl,4,4'-bis n,n-dimethylamino biphenyl,n,n,n',n'-tetramethyl-p,p'-benzidine,ccris 1000,1,1'-biphenyl-4,4'-diamine, n,n,n',n'-tetramethyl,n,n,n',n'-tetramethyl-1,1'-biphenyl-4,4'-diamine,4-4-dimethylamino phenyl phenyl dimethylamine,1,1'-biphenyl-4,4'-diamine, n4,n4,n4',n4'-tetramethyl
9702
4-[4-(dimethylamino)phenyl]-N,N-dimethylaniline
CN(C)C1=CC=C(C=C1)C2=CC=C(C=C2)N(C)C
Packaging Glass bottle
Quantity 25g
Infrared Spectrum Authentic
Melting Point 192°C to 196°C
Color Brown
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97.50%
Linear Formula ((CH3)2NC6H4-)2
Beilstein 13, 221
Formula Weight 240.35
Physical Form Fine Powder
Percent Purity ≥96.5%
Chemical Name or Material N,N,N',N'-Tetramethylbenzidine, 97.5%
Show More Show Less

EINECSNumber : 206-676-5

RTECSNumber : DD1752100

TSCA : TSCA