Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,N',N'-Tetramethylethylenediamine 99%, ACROS Organics™

Click to view available options
Quantity:
50mL
500mL
2.5L
Packaging:
Glass bottle
110-18-9
C6H16N2
116.21
KWYHDKDOAIKMQN-UHFFFAOYSA-N
temed,n,n,n',n'-tetramethylethylenediamine,tmeda,1,2-bis dimethylamino ethane,tetramethylethylenediamine,tetramethyldiaminoethane,tetrameen,propamine d,n1,n1,n2,n2-tetramethylethane-1,2-diamine,1,2-ethanediamine, n,n,n',n'-tetramethyl
8037
CHEBI:32850
N,N,N',N'-tetramethylethane-1,2-diamine
CN(C)CCN(C)C
110-18-9
C6H16N2
116.21
KWYHDKDOAIKMQN-UHFFFAOYSA-N
temed,n,n,n',n'-tetramethylethylenediamine,tmeda,1,2-bis dimethylamino ethane,tetramethylethylenediamine,tetramethyldiaminoethane,tetrameen,propamine d,n1,n1,n2,n2-tetramethylethane-1,2-diamine,1,2-ethanediamine, n,n,n',n'-tetramethyl
8037
CHEBI:32850
N,N,N',N'-tetramethylethane-1,2-diamine
CN(C)CCN(C)C
Packaging Glass bottle
Water 1% Max. (K.F.)
Refractive Index 1.4169 to 1.4189
Boiling Point 120.0°C to 122.0°C
Infrared Spectrum Authentic
Specific Gravity 0.77
Flash Point 17°C
Quantity 500mL
Color Undesignated
Melting Point -55.0°C
Density 0.7700g/mL
Assay Percent Range 99%
Linear Formula (CH3)2NCH2CH2N(CH3)2
Beilstein 04,25
Fieser 02,403; 03,284; 04,485; 05,652; 06,576; 07,358; 11,340; 12,477; 14,68; 15,63; 16,55
Merck Index 15,9277
Solubility Information Solubility in water: miscible. Other solubilities: most organic solvents
Formula Weight 116.21
Physical Form Liquid
Percent Purity 99%
Grade Extra Pure
Chemical Name or Material N,N,N',N'-Tetramethylethylenediamine
Show More Show Less