Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,N′,N′-Tetrapentylmalonamide 98.0+%, TCI America™

Click to view available options
Quantity:
5g
1560797-76-3
C23H46N2O2
382.633
MFCD06797172
SLCSFXNOZDNIFH-UHFFFAOYSA-N
N,N,N′C,N′C-Tetraamylmalonamide
44630524
N,N,N',N'-tetrapentylpropanediamide
CCCCCN(CCCCC)C(=O)CC(=O)N(CCCCC)CCCCC
1560797-76-3
C23H46N2O2
382.633
MFCD06797172
SLCSFXNOZDNIFH-UHFFFAOYSA-N
N,N,N′C,N′C-Tetraamylmalonamide
44630524
N,N,N',N'-tetrapentylpropanediamide
CCCCCN(CCCCC)C(=O)CC(=O)N(CCCCC)CCCCC
Color Yellow
Quantity 5g
Formula Weight 382.63
Physical Form Clear Liquid at 20°C
Chemical Name or Material N,N,N′,N′-Tetrapentylmalonamide