Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,N′-Trimethyl-N′-(2-hydroxyethyl)bis(2-aminoethyl) Ether 98.0+%, TCI America™

Click to view available options
Quantity:
25mL
500mL
83016-70-0
C9H22N2O2
190.287
MFCD00671578
NCUPDIHWMQEDPR-UHFFFAOYSA-N
N-(2-Hydroxyethyl)-N,N′,N′-trimethylbis(2-aminoethyl) Ether, 2-[[2-[2-(Dimethylamino)ethoxy]ethyl]methylamino]ethanol
3086157
2-[2-[2-(dimethylamino)ethoxy]ethyl-methylamino]ethanol
CN(C)CCOCCN(C)CCO
83016-70-0
C9H22N2O2
190.287
MFCD00671578
NCUPDIHWMQEDPR-UHFFFAOYSA-N
N-(2-Hydroxyethyl)-N,N′,N′-trimethylbis(2-aminoethyl) Ether, 2-[[2-[2-(Dimethylamino)ethoxy]ethyl]methylamino]ethanol
3086157
2-[2-[2-(dimethylamino)ethoxy]ethyl-methylamino]ethanol
CN(C)CCOCCN(C)CCO
Color Yellow
Quantity 500mL
UN Number 2735
Formula Weight 190.29
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material N,N,N′-Trimethyl-N′-(2-hydroxyethyl)bis(2-aminoethyl) Ether

EINECSNumber : (2)-4166

TSCA : Yes