Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,N'-Trimethylethylenediamine, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
142-25-6
C5H14N2
102.18
MFCD00014874
HVOYZOQVDYHUPF-UHFFFAOYSA-N
n,n,n'-trimethylethylenediamine,n1,n1,n2-trimethylethane-1,2-diamine,1,2-ethanediamine, n,n,n'-trimethyl,2-dimethylamino ethyl methyl amine,n,n,n-trimethylethylenediamine,ethylenediamine, n,n,n'-trimethyl,n1,n1,n2-trimethyl-1,2-ethanediamine,1,2-ethanediamine, n1,n1,n2-trimethyl,dimethyl 2-methylamino ethyl amine,n,n,n'-trimethylethane-1,2-diamine
67338
N,N',N'-trimethylethane-1,2-diamine
CNCCN(C)C
142-25-6
C5H14N2
102.18
MFCD00014874
HVOYZOQVDYHUPF-UHFFFAOYSA-N
n,n,n'-trimethylethylenediamine,n1,n1,n2-trimethylethane-1,2-diamine,1,2-ethanediamine, n,n,n'-trimethyl,2-dimethylamino ethyl methyl amine,n,n,n-trimethylethylenediamine,ethylenediamine, n,n,n'-trimethyl,n1,n1,n2-trimethyl-1,2-ethanediamine,1,2-ethanediamine, n1,n1,n2-trimethyl,dimethyl 2-methylamino ethyl amine,n,n,n'-trimethylethane-1,2-diamine
67338
N,N',N'-trimethylethane-1,2-diamine
CNCCN(C)C
Boiling Point 116°C to 118°C
Flash Point 9°C
Density 0.8000g/mL
Packaging Glass bottle
Specific Gravity 0.8
Refractive Index 1.4185 to 1.4205
Quantity 5g
Color Colorless
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula (CH3)2NCH2CH2NHCH3
Beilstein 04, I, 415
Formula Weight 102.18
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material N, N, N'-Trimethylethylenediamine
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful in contact with skin.
Causes serious eye damage.
Highly flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

EINECSNumber : 205-529-2