Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N,N,N'-Trimethylethylenediamine, 97%, ACROS Organics™
Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
Specifications
Density | 0.8000g/mL |
Boiling Point | 116°C to 118°C |
Refractive Index | 1.4185 to 1.4205 |
Flash Point | 9°C |
Specific Gravity | 0.8 |
Quantity | 25g |
Color | Colorless |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
CAS Min % | 96.0 |
Show More |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful in contact with skin.
Causes serious eye damage.
Highly flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
GHS Signal Word: Danger
EINECSNumber : 205-529-2