Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N,N',N''-Triphenylguanidine, 97%

Click to view available options
Quantity:
5g
25g
100g
101-01-9
C19H17N3
287.366
MFCD00020648
FUPAJKKAHDLPAZ-UHFFFAOYSA-N
n,n',n-triphenylguanidine,guanidine, n,n',n-triphenyl,sym-triphenylguanidine,guanidine, triphenyl,1,2,3-trifenylguanidin,unii-i64b170qfg,1,2,3-trifenylguanidin czech,guanidine, 1,2,3-triphenyl,phenyl 2-phenyl-1-phenylamino-2-azavinyl amine,triphenylguanidin
7539
1,2,3-triphenylguanidine
C1=CC=C(C=C1)NC(=NC2=CC=CC=C2)NC3=CC=CC=C3
101-01-9
C19H17N3
287.366
MFCD00020648
FUPAJKKAHDLPAZ-UHFFFAOYSA-N
n,n',n-triphenylguanidine,guanidine, n,n',n-triphenyl,sym-triphenylguanidine,guanidine, triphenyl,1,2,3-trifenylguanidin,unii-i64b170qfg,1,2,3-trifenylguanidin czech,guanidine, 1,2,3-triphenyl,phenyl 2-phenyl-1-phenylamino-2-azavinyl amine,triphenylguanidin
7539
1,2,3-triphenylguanidine
C1=CC=C(C=C1)NC(=NC2=CC=CC=C2)NC3=CC=CC=C3
Applications
It is employed as a intermediate for organic synthesis.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Melting Point 142°C to 145°C
Quantity 5g
Solubility Information Slightly soluble in water.
Formula Weight 287.36
Percent Purity 97%
Chemical Name or Material N,N',N''-Triphenylguanidine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 202-907-9

RTECSNumber : MF6825000

TSCA : Yes

Recommended Storage : Ambient temperatures