Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-p-Phenylenedimaleimide, ACROS Organics™

Click to view available options
Quantity:
10g
Packaging:
Glass bottle
3278-31-7
C14H8N2O4
268.228
MFCD00005505
AQGZJQNZNONGKY-UHFFFAOYSA-N
n,n'-1,4-phenylenedimaleimide,n,n'-p-phenylenedimaleimide,n,n'-4-phenylenedimaleimide,phenyl-1,4-bismaleimide,unii-bec7p1e6j1,1,1'-1,4-phenylene bis 1h-pyrrole-2,5-dione,bec7p1e6j1,1-4-2,5-dioxopyrrol-1-yl phenyl pyrrole-2,5-dione,p-pdm,1,4-dimaleimidobenzene
76765
1-[4-(2,5-dioxopyrrol-1-yl)phenyl]pyrrole-2,5-dione
C1=CC(=CC=C1N2C(=O)C=CC2=O)N3C(=O)C=CC3=O
3278-31-7
C14H8N2O4
268.228
MFCD00005505
AQGZJQNZNONGKY-UHFFFAOYSA-N
n,n'-1,4-phenylenedimaleimide,n,n'-p-phenylenedimaleimide,n,n'-4-phenylenedimaleimide,phenyl-1,4-bismaleimide,unii-bec7p1e6j1,1,1'-1,4-phenylene bis 1h-pyrrole-2,5-dione,bec7p1e6j1,1-4-2,5-dioxopyrrol-1-yl phenyl pyrrole-2,5-dione,p-pdm,1,4-dimaleimidobenzene
76765
1-[4-(2,5-dioxopyrrol-1-yl)phenyl]pyrrole-2,5-dione
C1=CC(=CC=C1N2C(=O)C=CC2=O)N3C(=O)C=CC3=O
Color Yellow
Melting Point >300°C (decomposition)
Quantity 10g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 268.23
Physical Form Powder
Percent Purity ≥98.0%
Chemical Name or Material N, N';-p-Phenylenedimaleimide
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Fatal if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger