N,Nft.-Bis(acryloyl)cystamine, 98%, Alfa Aesar™

Manufacturer: Alfa Aesar 4413206

Quantity 5g

 View more versions of this product

Catalog No. AA4413206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 60984-57-8
    Physical Form Powder, Packaged Under Argon
    Assay Percent Range 98%
    Molecular Formula C10H16N2O2S2
    Formula Weight 260.37
    MDL Number MFCD00036225
    Chemical Name or Material N, N′-Bis(acryloyl)cystamine

    Specifications

    Physical Form Powder, Packaged Under Argon
    Formula Weight 260.37
    Quantity 5g
    EINECS Number 262-546-8
    Assay Percent Range 98%
    Molecular Formula C10H16N2O2S2
    MDL Number MFCD00036225
    CAS 60984-57-8
    Chemical Name or Material N, N′-Bis(acryloyl)cystamine

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit