Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Nonanoic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
1kg
2.5kg
Packaging:
Glass bottle
112-05-0
C9H18O2
158.24
MFCD00004433
FBUKVWPVBMHYJY-UHFFFAOYSA-N
pelargonic acid,n-nonanoic acid,nonylic acid,pelargic acid,nonoic acid,n-nonylic acid,n-nonoic acid,1-octanecarboxylic acid,pelargon,cirrasol 185a
8158
CHEBI:29019
nonanoic acid
CCCCCCCCC(=O)O
112-05-0
C9H18O2
158.24
MFCD00004433
FBUKVWPVBMHYJY-UHFFFAOYSA-N
pelargonic acid,n-nonanoic acid,nonylic acid,pelargic acid,nonoic acid,n-nonylic acid,n-nonoic acid,1-octanecarboxylic acid,pelargon,cirrasol 185a
8158
CHEBI:29019
nonanoic acid
CCCCCCCCC(=O)O
Specific Gravity 0.9
Quantity 2.5kg
Refractive Index 1.4299 to 1.4339
Infrared Spectrum Authentic
Flash Point 114°C
Packaging Glass bottle
Color Colorless
Density 0.9000g/mL
Boiling Point 254.0°C
Melting Point 12.5°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula CH3(CH2)7CO2H
Beilstein 02, 352
Merck Index 15, 7179
Solubility Information Solubility in water: negligible. Other solubilities: soluble in alcohol, chloroform and ether, soluble in most other commmon organic solvents
Formula Weight 158.24
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material n-Nonanoic acid
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protec

GHS Signal Word: Warning

EINECSNumber : 203-931-2