Alfa Aesar™ N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% TMCS, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer: Alfa Aesar™ 4392814

Quantity 25g

 View more versions of this product

Catalog No. AA4392814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 25561-30-2
    Molecular Formula C8H18F3NOSi2
    Formula Weight 257.4
    MDL Number MFCD00008269
    UN Number UN2924
    Beilstein 3607620
    Chemical Name or Material N,O-Bis (trimethylsilyl)trifluoroacetamide, With 1% TMCS

    Specifications

    Formula Weight 257.4
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Physical Form Liquid
    Refractive Index 1.3840
    Melting Point −10°C
    Quantity 25g
    Flash Point 23°C (73°F)
    Sensitivity Moisture Sensitive
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H226
    Causes severe skin burns and eye damage.
    Flammable liquid and vapor.
    Storage Note 1
    • Packaged under Argon
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
    Molecular Formula C8H18F3NOSi2
    UN Number UN2924
    Beilstein 3607620
    Recommended Storage 2°C to 8°C
    Boiling Point 145° to 147°C
    MDL Number MFCD00008269
    CAS 25561-30-2
    Density 0.97g/mL
    Color Colorless to pale yellow
    Packaging ChemSeal™ bottles
    Chemical Name or Material N,O-Bis (trimethylsilyl)trifluoroacetamide, With 1% TMCS

    GHS H Statement
    H314-H226
    Causes severe skin burns and eye damage.
    Flammable liquid and vapor.

    2°C to 8°C

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged under Argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit