Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% trimethylsilyl chloride, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
25561-30-2,75-77-4
C8H18F3NOSi2
257.4
MFCD00008269
XCOBLONWWXQEBS-GHXNOFRVSA-N
bstfa,n,o-bis trimethylsilyl trifluoroacetamide,acetamide, 2,2,2-trifluoro-o,n-bis trimethylsilyl,trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl acetimidate,trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanimidate,trimethylsilyl 1z-2,2,2-trifluoro-n-z-trimethylsilyl ethanimidoate #,n,o-bis trimethylsilyl trifluoroacetamide bstfa,trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanecarboximidate
9601896
trimethylsilyl (1Z)-2,2,2-trifluoro-N-trimethylsilylethanimidate
C[Si](C)(C)N=C(C(F)(F)F)O[Si](C)(C)C
25561-30-2,75-77-4
C8H18F3NOSi2
257.4
MFCD00008269
XCOBLONWWXQEBS-GHXNOFRVSA-N
bstfa,n,o-bis trimethylsilyl trifluoroacetamide,acetamide, 2,2,2-trifluoro-o,n-bis trimethylsilyl,trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl acetimidate,trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanimidate,trimethylsilyl 1z-2,2,2-trifluoro-n-z-trimethylsilyl ethanimidoate #,n,o-bis trimethylsilyl trifluoroacetamide bstfa,trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanecarboximidate
9601896
trimethylsilyl (1Z)-2,2,2-trifluoro-N-trimethylsilylethanimidate
C[Si](C)(C)N=C(C(F)(F)F)O[Si](C)(C)C
Silylation reagent
Specific Gravity 0.974
Quantity 500g
Boiling Point 145.0°C to 147.0°C
Density 0.9740g/mL
Flash Point 34°C
Color Colorless to Yellow
Linear Formula CF3C[=NSi(CH3)3]OSi(CH3)3
Solubility Information Solubility in water: hydrolysis
Formula Weight 257.4
Physical Form Liquid
Percent Purity ≥98.0% (excluding TMCS), 1.5% max. TMCS
Chemical Name or Material N, O-Bis(trimethylsilyl)trifluoroacetamide
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 247-103-9