Alfa Aesar™ N,O-Bis(trimethylsilyl)trifluoroacetamide, with 10% TMCS

Manufacturer: Alfa Aesar™ 4017422

Quantity 100g

 View more versions of this product

Catalog No. AA4017422

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 25561-30-2
    Formula Weight 257.4
    MDL Number MFCD00008269
    UN Number UN2924
    Beilstein 3607620
    Chemical Name or Material N,O-Bis(trimethylsilyl)trifluoroacetamide, with 10% TMCS

    Specifications

    Formula Weight 257.4
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 1 Danger
    Physical Form Liquid
    Beilstein 3607620
    Storage Note 1 2°C to 8°C
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
    Sensitivity Moisture Sensitive
    MDL Number MFCD00008269
    UN Number UN2924
    TSCA TSCA
    Color Colorless to pale yellow
    Flash Point 23° (73°F)
    CAS 25561-30-2
    Health Hazard 2 GHS H Statement
    H225-H314
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Quantity 100g
    Chemical Name or Material N,O-Bis(trimethylsilyl)trifluoroacetamide, with 10% TMCS

    GHS H Statement
    H225-H314
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    2°C to 8°C

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit