Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,O-Dimethylhydroxylamine Hydrochloride 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
6638-79-5
C2H7NO·HCl
97.55
MFCD00012485
USZLCYNVCCDPLQ-UHFFFAOYSA-N
n,o-dimethylhydroxylamine hydrochloride,n,o-dimethylhydroxylamine hcl,n-methoxymethylamine hydrochloride,methanamine, n-methoxy-, hydrochloride,o,n-dimethylhydroxylamine hydrochloride,n-methoxymethanamine hydrochloride,o,n-dimethyl-hydroxylamine hydrochloride,methoxy methyl amine hydrochloride,n,o-dimethylhydroxyamine hydrochloride,n,o-dimethylhydroxylaminehydrochloride
81138
N-methoxymethanamine;hydrochloride
CNOC.Cl
6638-79-5
C2H7NO·HCl
97.55
MFCD00012485
USZLCYNVCCDPLQ-UHFFFAOYSA-N
n,o-dimethylhydroxylamine hydrochloride,n,o-dimethylhydroxylamine hcl,n-methoxymethylamine hydrochloride,methanamine, n-methoxy-, hydrochloride,o,n-dimethylhydroxylamine hydrochloride,n-methoxymethanamine hydrochloride,o,n-dimethyl-hydroxylamine hydrochloride,methoxy methyl amine hydrochloride,n,o-dimethylhydroxyamine hydrochloride,n,o-dimethylhydroxylaminehydrochloride
81138
N-methoxymethanamine;hydrochloride
CNOC.Cl
Coupling reagent
Melting Point 112.0°C to 116.0°C
Water 0.5% max.
Packaging Glass bottle
Color White
Infrared Spectrum Authentic
Quantity 25g
Appearance (10% aq. soln.) clear
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3ONHCH3·HCl
Beilstein 04, 534
Fieser 11,201; 12,205; 15,144
Solubility Information Solubility in water: soluble
Formula Weight 97.55
Physical Form Crystalline Powder or Crystals
Percent Purity 98%
Chemical Name or Material N, O-Dimethylhydroxylamine hydrochloride, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC

EINECSNumber : 229-642-1

TSCA : TSCA